Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021296.hg.1 |
TC04001504.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021296.hg.1 |
ENST00000264805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021296.hg.1 |
ENST00000354960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021296.hg.1 |
ENST00000394439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021296.hg.1 |
NM_001083.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021296.hg.1 |
NM_033430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021296.hg.1 |
NM_033437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000264805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000354960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000394439.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000503412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000512494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
NM_001083.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
NM_033430.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
NM_033437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000264805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000354960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000394439.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000503412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
ENST00000512494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
NM_001083.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
NM_033430.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
NM_033437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180491.hg.1 |
TC0400011712.hg.1 |
Clariom D TC |