Assay Details
| Gene Symbol: SLC9A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042537.1 | NP_001036002.1 | 2 - 3 | 602 | 144 | |
| NM_001177651.1 | NP_001171122.1 | 3 - 4 | 480 | 144 | ||
| XM_006724726.3 | XP_006724789.1 | 3 - 4 | 596 | 144 | ||
| XM_017029223.1 | XP_016884712.1 | 3 - 4 | 613 | 144 | ||
| XM_017029224.1 | XP_016884713.1 | 3 - 4 | 743 | 144 | ||
| GenBank mRNA | AB074255.1 | - | 2 - 3 | 526 | 144 | |
| AK289775.1 | - | 2 - 3 | 526 | 144 | ||
| AK296444.1 | - | 2 - 3 | 265 | 144 | ||
| AK300475.1 | - | 3 - 4 | 471 | 144 | ||
| AK314562.1 | - | 2 - 3 | 594 | 144 | ||
| AK316255.1 | - | 3 - 4 | 471 | 144 | ||
| BC035029.1 | - | 2 - 3 | 564 | 5264865 | 144 | |
| BC049169.1 | - | 2 - 3 | 526 | 5312949 | 144 | |
| DC308430.1 | - | 3 - 4 | 481 | 144 | ||
More Information
Set Membership: |
|

SLC9A6
Genomic Map