Assay Details
| Gene Symbol: MTHFSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159377.1 | NP_001152849.1 | 3 - 4 | 288 | 87 | |
| NM_001159380.1 | NP_001152852.1 | 3 - 4 | 252 | 87 | ||
| NM_022764.2 | NP_073601.2 | 3 - 4 | 285 | 87 | ||
| XM_006721247.3 | XP_006721310.1 | 2 - 3 | 159 | 87 | ||
| XM_011523284.2 | XP_011521586.1 | 3 - 4 | 288 | 87 | ||
| XM_011523286.1 | XP_011521588.1 | 3 - 4 | 349 | 87 | ||
| XM_011523287.1 | XP_011521589.1 | 3 - 4 | 253 | 87 | ||
| XM_011523288.1 | XP_011521590.1 | 2 - 3 | 181 | 87 | ||
| XM_011523289.1 | XP_011521591.1 | 3 - 4 | 349 | 87 | ||
| XM_017023573.1 | XP_016879062.1 | 3 - 4 | 242 | 87 | ||
| GenBank mRNA | AK023060.1 | - | 3 - 4 | 263 | 87 | |
| AK023955.1 | - | 3 - 4 | 242 | 87 | ||
| BC013911.2 | - | 2 - 3 | 219 | 87 | ||
| BC112057.1 | - | 3 - 4 | 268 | 8327525 | 87 | |
| BC112059.1 | - | 3 - 4 | 268 | 8327527 | 87 | |
| BC143704.1 | - | 3 - 4 | 253 | 87 | ||
| BC143705.1 | - | 3 - 4 | 252 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006721247.3, XM_011523286.1, XM_011523287.1, XM_011523288.1, XM_011523289.1, XM_017023573.1 transcripts.
Set Membership: |
|

MTHFSD
Genomic Map