| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008181.hg.1 |
TC0X000620.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008181.hg.1 |
ENST00000218147.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008181.hg.1 |
ENST00000303743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008181.hg.1 |
ENST00000359304.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008181.hg.1 |
ENST00000488135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008181.hg.1 |
ENST00000540052.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008181.hg.1 |
NM_021946.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008182.hg.1 |
TC0X000620.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008182.hg.1 |
ENST00000218147.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008182.hg.1 |
ENST00000303743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008182.hg.1 |
ENST00000359304.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008182.hg.1 |
ENST00000488135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008182.hg.1 |
ENST00000540052.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008182.hg.1 |
NM_021946.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008185.hg.1 |
TC0X000620.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008185.hg.1 |
ENST00000218147.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008185.hg.1 |
ENST00000303743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008185.hg.1 |
ENST00000359304.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008185.hg.1 |
ENST00000488135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008185.hg.1 |
ENST00000540052.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008185.hg.1 |
NM_021946.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
ENST00000218147.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
ENST00000488135.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
ENST00000540052.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
ENST00000607874.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
NM_001184772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
NM_021946.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
ENST00000218147.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
ENST00000488135.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
ENST00000540052.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
ENST00000607874.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
NM_001184772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
NM_021946.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157170.hg.1 |
TC0X00008394.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
ENST00000218147.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
ENST00000488135.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
ENST00000540052.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
ENST00000607874.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
NM_001184772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
NM_021946.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
ENST00000218147.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
ENST00000488135.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
ENST00000540052.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
ENST00000607874.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
NM_001184772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
NM_021946.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157172.hg.1 |
TC0X00008394.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
ENST00000218147.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
ENST00000488135.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
ENST00000540052.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
NM_001184772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
NM_021946.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
ENST00000218147.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
ENST00000488135.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
ENST00000540052.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
NM_001184772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
NM_021946.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157175.hg.1 |
TC0X00008394.hg.1 |
Clariom D TC |