Assay Details
| Gene Symbol: BCORL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017029721.1 | XP_016885210.1 | 2 - 3 | 444 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017029721.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000441294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000441294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000441294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000441294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000218147.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000441294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000441294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000441294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000441294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000456822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000488135.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000540052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | ENST00000607874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_001184772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008394.hg.1 | NM_021946.4 | Refseq |

BCORL1
Genomic Map