Assay Details
| Gene Symbol: PIGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005264369.2 | XP_005264426.1 | 5 - 6 | 643 | 74 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |

PIGF
Genomic Map