Assay Details
| Gene Symbol: PIGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173074.2 | NP_775097.1 | 6 - 7 | 816 | 93 | |
| XM_011532908.2 | XP_011531210.1 | 6 - 7 | 698 | 93 | ||
| GenBank mRNA | BC021725.1 | - | 6 - 7 | 756 | 5269689 | 93 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001816.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02029066.hg.1 | TC02001816.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029066.hg.1 | ENST00000281382.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029066.hg.1 | ENST00000306465.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029066.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02029066.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191128.hg.1 | TC0200012489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200191131.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191131.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191131.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191131.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191131.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191131.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191131.hg.1 | TC0200012489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191129.hg.1 | TC0200012489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000281382.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000306465.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000412717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000420164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000474980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000482786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | ENST00000495933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_002643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012489.hg.1 | NM_173074.2 | Refseq |

PIGF
Genomic Map