Assay Details
| Gene Symbol: SLC9A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004174.3 | NP_004165.2 | 7 - 8 | 1470 | 66 | |
| GenBank mRNA | BC011555.1 | - | 5 - 6 | 1005 | 66 | |
| BC101669.1 | - | 7 - 8 | 1366 | 8069175 | 66 | |
| BC101671.1 | - | 7 - 8 | 1366 | 8069177 | 66 | |
| BP270851.1 | - | 1 - 2 | 101 | 66 | ||
| S83545.1 | - | 2 - 3 | 311 | 66 | ||
| U28043.1 | - | 7 - 8 | 1368 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001096.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of sodium ion transport
ion transport
receptor-mediated endocytosis
circadian rhythm
response to glucocorticoid
regulation of intracellular pH
potassium ion transmembrane transport
sodium ion import across plasma membrane
hydrogen ion transmembrane transport
protein binding
sodium:proton antiporter activity
potassium:proton antiporter activity
PDZ domain binding
regulation of sodium ion transport
ion transport
receptor-mediated endocytosis
circadian rhythm
response to glucocorticoid
regulation of intracellular pH
potassium ion transmembrane transport
sodium ion import across plasma membrane
hydrogen ion transmembrane transport
protein binding
sodium:proton antiporter activity
potassium:proton antiporter activity
PDZ domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05015825.hg.1 | TC05001096.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015825.hg.1 | ENST00000264938.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015825.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015824.hg.1 | TC05001096.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015824.hg.1 | ENST00000264938.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015824.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015824.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009805.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169082.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169082.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169082.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169082.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169082.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169082.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169082.hg.1 | TC0500009805.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | ENST00000264938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | ENST00000507407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | ENST00000514375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | NM_001284351.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | NM_004174.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169081.hg.1 | TC0500009805.hg.1 | Clariom D TC |

SLC9A3
Genomic Map