Assay Details
| Gene Symbol: CENPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042426.1 | NP_001035891.1 | 2 - 3 | 389 | 78 | |
| GenBank mRNA | BC000881.1 | - | 2 - 3 | 336 | 3461992 | 78 |
More Information
Set Membership: |
|

CENPA
Genomic Map