Assay Details
| Gene Symbol: LOC100129434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125368.1 | 3 | 1020 | 85 | ||
| GenBank mRNA | AK096196.1 | - | 3 | 1232 | 85 | |
| AK311113.1 | - | 3 | 1020 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P324054 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 34080 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P324054 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13219698 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8052379 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8052377 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8052378 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8052376 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000447423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012666.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | ENST00000432793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192457.hg.1 | TC0200012666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200192456.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192456.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192456.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192456.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192456.hg.1 | ENST00000596663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192456.hg.1 | NR_125368.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192456.hg.1 | TC0200012666.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3273185 | AK311113.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3273185 | AK096196.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3183896 | AK311113.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3183896 | AK096196.1 | human_transcriptome |

LOC100129434
Genomic Map