Assay Details
| Gene Symbol: EFNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182690.2 | NP_872632.2 | 3 - 4 | 551 | 140 | |
| GenBank mRNA | AJ006353.1 | - | 3 - 4 | 491 | 140 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006332.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01019878.hg.1 | TC01006332.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019878.hg.1 | ENST00000359751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019878.hg.1 | ENST00000368409.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019878.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019878.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019878.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019878.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019875.hg.1 | TC01006332.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019875.hg.1 | ENST00000359751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019875.hg.1 | ENST00000368409.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019875.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019875.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019875.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019875.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019877.hg.1 | TC01006332.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019877.hg.1 | ENST00000359751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019877.hg.1 | ENST00000368409.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019877.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019877.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019877.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019877.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239690.hg.1 | TC0100018302.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239691.hg.1 | TC0100018302.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239692.hg.1 | TC0100018302.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018302.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | ENST00000359751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | ENST00000368409.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | ENST00000427683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | NM_005227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | NM_182689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | NM_182690.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239693.hg.1 | TC0100018302.hg.1 | Clariom D TC |

EFNA4
Genomic Map