Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000319033.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000319033.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000319033.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000476832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000476832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000476832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
ENST00000476832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017603.hg.1 |
TC09001411.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017603.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000319033.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000319033.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000319033.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000476832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000476832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000476832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
ENST00000476832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NM_033087.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010995.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174941.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174941.hg.1 |
ENST00000319033.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174941.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174941.hg.1 |
ENST00000238477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174941.hg.1 |
ENST00000319033.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174941.hg.1 |
NR_024532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174941.hg.1 |
TC0900010995.hg.1 |
Clariom D TC |