Assay Details
Gene Symbol: BRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017028373.1 | XP_016883862.1 | 2 - 3 | 150 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017028373.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
