Assay Details
| Gene Symbol: SMARCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130420.1 | NP_001123892.1 | 18 - 19 | 1851 | 63 | |
| NM_139067.2 | NP_620706.1 | 18 - 19 | 1851 | 63 | ||
| XM_005269101.1 | XP_005269158.1 | 18 - 19 | 1788 | 63 | ||
| XM_011538693.2 | XP_011536995.1 | 17 - 18 | 1697 | 63 | ||
| GenBank mRNA | AB209006.1 | - | 18 - 19 | 1851 | 63 | |
| AK025128.1 | - | 3 - 4 | 276 | 63 | ||
| AK293896.1 | - | 18 - 19 | 1738 | 63 | ||
| BC013045.1 | - | 18 - 19 | 1777 | 4180388 | 63 | |
| BT009924.1 | - | 18 - 19 | 1745 | 63 | ||
More Information
Set Membership: |
|

SMARCC2
Genomic Map