Assay Details
| Gene Symbol: LINC00543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135254.1 | 1 | 780 | 68 | ||
| GenBank mRNA | AK000246.1 | - | 1 | 780 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit pseudogene 2 |
ATP5EP2 | -8.5kb | Anti-sense | Chr. 13 - 27945206 - 27945573 on NCBI Build GRCh38 | |
| Downstream | caudal type homeobox 2 | CDX2 |
CDX-3, CDX2/AS, CDX3 |
5.7kb | Sense | Chr. 13 - 27960918 - 27969368 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC00543
Genomic Map