Assay Details
| Gene Symbol: HERC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278186.1 | NP_001265115.1 | 14 - 15 | 1979 | 95 | |
| NM_015601.3 | NP_056416.2 | 16 - 17 | 2231 | 95 | ||
| XM_011539593.2 | XP_011537895.1 | 17 - 18 | 2305 | 95 | ||
| GenBank mRNA | AK027835.1 | - | 7 - 8 | 798 | 95 | |
| AK292809.1 | - | 16 - 17 | 2111 | 95 | ||
| AY650032.1 | - | 14 - 15 | 1916 | 95 | ||
| BC015043.1 | - | 14 - 15 | 1882 | 95 | ||
| BC039600.1 | - | 16 - 17 | 2141 | 5212048 | 95 | |
| BX537563.1 | - | 16 - 17 | 4718 | 95 | ||
More Information
Set Membership: |
|

HERC4
Genomic Map