Assay Details
| Gene Symbol: PABPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003819.3 | NP_003810.1 | 10 - 11 | 2300 | 82 | |
| GenBank mRNA | U33818.1 | - | 10 - 11 | 1555 | 82 | |
| U75686.1 | - | 10 - 11 | 2196 | 82 | ||
More Information
Set Membership: |
|

PABPC4
Genomic Map