Assay Details
| Gene Symbol: DCTN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006571.3 | NP_006562.1 | 1 - 2 | 111 | 97 | |
| GenBank mRNA | AK314128.1 | - | 1 - 2 | 68 | 97 | |
| BC013175.1 | - | 1 - 2 | 55 | 3925231 | 97 | |
| BI562535.1 | - | 1 - 2 | 97 | 97 | ||
| D84145.1 | - | 1 - 2 | 111 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08002579.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK314128.1,BC013175.1,D84145.1,NM_006571.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08003376.hg.1 | TC08002579.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003376.hg.1 | ENST00000221114.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003376.hg.1 | ENST00000520829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003376.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003371.hg.1 | TC08002579.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003371.hg.1 | ENST00000221114.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003371.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003375.hg.1 | TC08002579.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003375.hg.1 | ENST00000221114.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003375.hg.1 | ENST00000520829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003375.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012288.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000519830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000520829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000522540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | ENST00000523666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185113.hg.1 | TC0800012288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800185107.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185107.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185107.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185107.hg.1 | ENST00000221114.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185107.hg.1 | ENST00000522141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185107.hg.1 | NM_006571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185107.hg.1 | TC0800012288.hg.1 | Clariom D TC |

DCTN6
Genomic Map