Assay Details
| Gene Symbol: TAX1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079864.2 | NP_001073333.1 | 13 - 14 | 1933 | 132 | |
| NM_001206901.1 | NP_001193830.1 | 13 - 14 | 1936 | 132 | ||
| NM_001206902.1 | NP_001193831.1 | 12 - 13 | 1745 | 132 | ||
| GenBank mRNA | AF090891.1 | - | 13 - 14 | 1851 | 132 | |
| AK296720.1 | - | 12 - 13 | 1694 | 132 | ||
| AK299729.1 | - | 13 - 14 | 1936 | 132 | ||
| AK301064.1 | - | 13 - 14 | 1722 | 132 | ||
| AK309757.1 | - | 2 - 3 | 147 | 132 | ||
| CR457056.1 | - | 12 - 13 | 1751 | 132 | ||
More Information
Set Membership: |
|

TAX1BP1
Genomic Map