Assay Details
| Gene Symbol: AOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277731.1 | NP_001264660.1 | 1 - 2 | 1759 | 58 | |
| NM_003734.3 | NP_003725.1 | 1 - 2 | 1759 | 58 | ||
| GenBank mRNA | AF067406.1 | - | 1 - 2 | 1679 | 58 | |
| AK315129.1 | - | 1 - 2 | 1738 | 58 | ||
| BC050549.1 | - | 1 - 2 | 1709 | 6193046 | 58 | |
| JX020506.1 | - | 1 - 2 | 1687 | 58 | ||
| U39447.1 | - | 1 - 2 | 1759 | 58 | ||
More Information
Set Membership: |
|

AOC3
Genomic Map