Assay Details
| Gene Symbol: PBK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018492.3 | NP_060962.2 | 1 - 2 | 445 | 70 | |
| GenBank mRNA | AB027249.1 | - | 1 - 2 | 171 | 70 | |
| AB027250.1 | - | 1 - 2 | 151 | 70 | ||
| AF237709.1 | - | 1 - 2 | 159 | 70 | ||
| AK027291.1 | - | 1 - 2 | 150 | 70 | ||
| BC015191.1 | - | 1 - 2 | 159 | 4082846 | 70 | |
| BI461026.1 | - | 1 - 2 | 188 | 70 | ||
| DB028500.1 | - | 1 - 2 | 445 | 70 | ||
| EU794685.1 | - | 1 - 2 | 182 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001086.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB027249.1,AB027250.1,AF237709.1,AK027291.1,BC015191.1,EU794685.1,NM_018492.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
mitotic nuclear division
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of stress-activated MAPK cascade
cellular response to UV
negative regulation of inflammatory response
protein serine/threonine kinase activity
protein binding
ATP binding
protein phosphorylation
mitotic nuclear division
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of stress-activated MAPK cascade
cellular response to UV
negative regulation of inflammatory response
protein serine/threonine kinase activity
protein binding
ATP binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08013943.hg.1 | TC08001086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013943.hg.1 | ENST00000301905.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013943.hg.1 | ENST00000522944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013943.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013947.hg.1 | TC08001086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013947.hg.1 | ENST00000301905.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013947.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013948.hg.1 | TC08001086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013948.hg.1 | ENST00000301905.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013948.hg.1 | ENST00000522944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013948.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013946.hg.1 | TC08001086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013946.hg.1 | ENST00000301905.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013946.hg.1 | ENST00000522944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013946.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013944.hg.1 | TC08001086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013944.hg.1 | ENST00000301905.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013944.hg.1 | ENST00000522944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013944.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013949.hg.1 | TC08001086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013949.hg.1 | ENST00000301905.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013949.hg.1 | ENST00000522944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013949.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167805.hg.1 | TC0800009970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800167807.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167807.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167807.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167807.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167807.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167807.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167807.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167807.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167807.hg.1 | TC0800009970.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 169723 | - | - |

PBK
Genomic Map