Assay Details
| Gene Symbol: CCRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003965.4 | NP_003956.2 | 1 - 2 | 278 | 134 | |
| XM_011534208.1 | XP_011532510.1 | 2 - 3 | 222 | 134 | ||
| XM_011534209.1 | XP_011532511.1 | 2 - 3 | 145 | 134 | ||
| GenBank mRNA | AF015525.1 | - | 1 - 2 | 320 | 134 | |
| BC071682.1 | - | 1 - 2 | 231 | 5182006 | 134 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000257.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF015525.1,BC071682.1,NM_003965.4,XM_011534208.1,XM_011534209.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03005217.hg.1 | TC03000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005217.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005217.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005217.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000441909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000441909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005216.hg.1 | TC03000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005216.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005216.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005215.hg.1 | TC03000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005215.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005215.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005214.hg.1 | TC03000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005214.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005214.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005218.hg.1 | TC03000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005218.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005218.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005218.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151436.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151436.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151436.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151436.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151436.hg.1 | TC0300007259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151438.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151438.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151438.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151438.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151438.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151438.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151438.hg.1 | TC0300007259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151439.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151439.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151439.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151439.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151439.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151439.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151439.hg.1 | TC0300007259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000357392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400880.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000400882.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000433848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000441909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000441909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | ENST00000495870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_001130910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007259.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151435.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151435.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151435.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151435.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151435.hg.1 | TC0300007259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151437.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151437.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151437.hg.1 | ENST00000399036.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151437.hg.1 | NM_003965.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151437.hg.1 | TC0300007259.hg.1 | Clariom D TC |

CCRL2
Genomic Map