Assay Details
| Gene Symbol: NCK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110177.1 | 1 - 2 | 310 | 115 | ||
| GenBank mRNA | DA714434.1 | - | 1 - 2 | 278 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012551.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193850.hg.1 | TC0300012551.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193855.hg.1 | TC0300012551.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000461864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000470236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | ENST00000474250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | NR_110175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | NR_110176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | NR_110177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193856.hg.1 | TC0300012551.hg.1 | Clariom D TC |

NCK1-AS1
Genomic Map