Assay Details
| Gene Symbol: LOC105371335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135171.1 | 1 - 2 | 100 | |||
| GenBank mRNA | HY237382.1 | - | 2 - 3 | 369 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371335| Gene Symbol: LOC105371335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135171.1 | 1 - 2 | 100 | |||
| GenBank mRNA | HY237382.1 | - | 2 - 3 | 369 | 100 | |
Set Membership: |
|