Assay Details
| Gene Symbol: SIPA1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020808.4 | NP_065859.3 | 12 - 13 | 3908 | 76 | |
| XM_005273211.2 | XP_005273268.1 | 14 - 15 | 4245 | 76 | ||
| XM_005273212.4 | XP_005273269.1 | 14 - 15 | 4052 | 76 | ||
| XM_005273213.4 | XP_005273270.1 | 13 - 14 | 3945 | 76 | ||
| XM_011544243.2 | XP_011542545.1 | 15 - 16 | 4101 | 76 | ||
| XM_017001896.1 | XP_016857385.1 | 14 - 15 | 3998 | 76 | ||
| XM_017001897.1 | XP_016857386.1 | 14 - 15 | 4245 | 76 | ||
| XM_017001898.1 | XP_016857387.1 | 14 - 15 | 4019 | 76 | ||
| GenBank mRNA | AB037810.1 | - | 12 - 13 | 3112 | 76 | |
| AK022852.1 | - | 4 - 5 | 508 | 76 | ||
| AK093191.1 | - | 6 - 7 | 1044 | 76 | ||
| AY168879.1 | - | 12 - 13 | 3868 | 76 | ||
| BC013119.1 | - | 4 - 5 | 419 | 76 | ||
| BC082251.1 | - | 4 - 5 | 448 | 76 | ||
| BC150334.1 | - | 12 - 13 | 3123 | 8860510 | 76 | |
More Information
Set Membership: |
|

SIPA1L2
Genomic Map