Assay Details
| Gene Symbol: FER1L5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011512110.2 | XP_011510412.1 | 46 - 47 | 5278 | 66 | |
| XM_011512111.2 | XP_011510413.1 | 46 - 47 | 5275 | 66 | ||
| XM_011512113.2 | XP_011510415.1 | 46 - 47 | 5272 | 66 | ||
| XM_011512115.2 | XP_011510417.1 | 45 - 46 | 5209 | 66 | ||
| XM_011512116.2 | XP_011510418.1 | 46 - 47 | 5191 | 66 | ||
| XM_011512117.2 | XP_011510419.1 | 45 - 46 | 5173 | 66 | ||
| XM_011512118.2 | XP_011510420.1 | 45 - 46 | 5158 | 66 | ||
| XM_011512119.2 | XP_011510421.1 | 45 - 46 | 5116 | 66 | ||
| XM_011512120.2 | XP_011510422.1 | 45 - 46 | 5169 | 66 | ||
| XM_011512122.2 | XP_011510424.1 | 46 - 47 | 5279 | 66 | ||
| XM_011512124.2 | XP_011510426.1 | 30 - 31 | 4862 | 66 | ||
| XM_017005222.1 | XP_016860711.1 | 46 - 47 | 5275 | 66 | ||
| XM_017005223.1 | XP_016860712.1 | 29 - 30 | 3926 | 66 | ||
| XM_017005224.1 | XP_016860713.1 | 20 - 21 | 2718 | 66 | ||
More Information
Set Membership: |
|

FER1L5
Genomic Map