Assay Details
| Gene Symbol: ARNTL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011520769.2 | XP_011519071.1 | 3 - 4 | 439 | 74 | |
| GenBank mRNA | AF256215.1 | - | 438 | 74 | ||
More Information
Set Membership: |
|

ARNTL2
Genomic Map