Assay Details
| Gene Symbol: B3GNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011513227.2 | XP_011511529.1 | 1 - 2 | 270 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011513227.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
central nervous system development
brain development
glycolipid biosynthetic process
O-glycan processing
beta-galactosyl-N-acetylglucosaminylgalactosylglucosyl-ceramide beta-1,3-acetylglucosaminyltransferase activity
UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
lactosylceramide 1,3-N-acetyl-beta-D-glucosaminyltransferase activity
protein glycosylation
central nervous system development
brain development
glycolipid biosynthetic process
O-glycan processing
beta-galactosyl-N-acetylglucosaminylgalactosylglucosyl-ceramide beta-1,3-acetylglucosaminyltransferase activity
UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
lactosylceramide 1,3-N-acetyl-beta-D-glucosaminyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000477699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000477699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000477699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000480551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000480551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000480551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000326505.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000460419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000462559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000464923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000465010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000477699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000477699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000477699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000480551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000480551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000480551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000481531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000488301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000493370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | ENST00000496270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009661.hg.1 | NM_032047.4 | Refseq |

B3GNT5
Genomic Map