Assay Details
| Gene Symbol: ORC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190879.2 | NP_001177808.1 | 2 - 3 | 271 | 117 | |
| XM_011511255.2 | XP_011509557.1 | 2 - 3 | 309 | 117 | ||
| GenBank mRNA | BX423253.2 | - | 2 - 3 | 249 | 117 | |
| DC302315.1 | - | 2 - 3 | 262 | 117 | ||
| DC399282.1 | - | 2 - 3 | 272 | 117 | ||
| DC399552.1 | - | 2 - 3 | 261 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190879.2, XM_011511255.2 transcripts.
Set Membership: |
|

ORC4
Genomic Map