Assay Details
| Gene Symbol: ORC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK299749.1 | - | 6 - 7 | 498 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK299749.1 transcripts.
Set Membership: |
|

ORC4
Genomic Map