Assay Details
| Gene Symbol: TGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201631.3 | NP_963925.2 | 3 - 4 | 443 | 112 | |
| GenBank mRNA | AF035960.1 | - | 3 - 4 | 443 | 112 | |
| BC119009.1 | - | 3 - 4 | 443 | 40085374 | 112 | |
| BC122859.1 | - | 3 - 4 | 443 | 40085372 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500167130.hg.1 | TC1500009214.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000220420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000349114.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000396996.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000563838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000610827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000611276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | ENST00000622115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_004245.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009214.hg.1 | NM_201631.3 | Refseq |

TGM5
Genomic Map