Assay Details
| Gene Symbol: SLC2A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104247.1 | 5 - 6 | 802 | 104 | ||
| GenBank mRNA | AB209301.1 | - | 4 - 5 | 104 | ||
| AK296169.1 | - | 5 - 6 | 571 | 104 | ||
| BC100807.2 | - | 5 - 6 | 104 | |||
| BC100810.1 | - | 5 - 6 | 548 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC100807.2, BC100810.1 transcripts.
Set Membership: |
|

SLC2A11
Genomic Map