Assay Details
| Gene Symbol: SLC28A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199633.1 | NP_001186562.1 | 16 - 17 | 1977 | 80 | |
| NM_022127.2 | NP_071410.1 | 17 - 18 | 1923 | 80 | ||
| NR_037638.2 | 17 - 18 | 2153 | 80 | |||
| XM_011518905.2 | XP_011517207.1 | 16 - 17 | 2047 | 80 | ||
| XM_011518906.2 | XP_011517208.1 | 17 - 18 | 2143 | 80 | ||
| XM_011518907.2 | XP_011517209.1 | 15 - 16 | 1862 | 80 | ||
| XM_011518908.2 | XP_011517210.1 | 11 - 12 | 1882 | 80 | ||
| GenBank mRNA | AF305210.1 | - | 17 - 18 | 1923 | 80 | |
| AK292849.1 | - | 16 - 17 | 1976 | 80 | ||
| AK304406.1 | - | 17 - 18 | 2133 | 80 | ||
| AK314039.1 | - | 16 - 17 | 1957 | 80 | ||
| BC093821.1 | - | 16 - 17 | 1860 | 7939666 | 80 | |
| BC093823.1 | - | 16 - 17 | 1860 | 7939668 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001286.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
retina homeostasis
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
pyrimidine-containing compound transmembrane transport
purine nucleobase transmembrane transport
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity
retina homeostasis
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
pyrimidine-containing compound transmembrane transport
purine nucleobase transmembrane transport
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016075.hg.1 | TC09001286.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016075.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016075.hg.1 | ENST00000537648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016075.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016075.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016075.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172271.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900172271.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172271.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172271.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172271.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900172271.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172271.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172271.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172271.hg.1 | TC0900010591.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |

SLC28A3
Genomic Map