Assay Details
| Gene Symbol: OLFM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006418.4 | NP_006409.3 | 3 - 4 | 662 | 80 | |
| GenBank mRNA | AF097021.1 | - | 3 - 4 | 579 | 80 | |
| AK300947.1 | - | 3 - 4 | 542 | 80 | ||
| AY358567.1 | - | 3 - 4 | 579 | 80 | ||
| BC047740.1 | - | 2 - 3 | 293 | 80 | ||
| BC117329.1 | - | 3 - 4 | 644 | 40125880 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

OLFM4
Genomic Map