Assay Details
| Gene Symbol: RANBP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022897.4 | NP_075048.1 | 5 - 6 | 636 | 100 | |
| XM_011534627.1 | XP_011532929.1 | 5 - 6 | 492 | 100 | ||
| XM_011534631.2 | XP_011532933.1 | 4 - 5 | 505 | 100 | ||
| XM_011534636.2 | XP_011532938.1 | 5 - 6 | 492 | 100 | ||
| XM_011534637.2 | XP_011532939.1 | 5 - 6 | 492 | 100 | ||
| XM_017009736.1 | XP_016865225.1 | 5 - 6 | 492 | 100 | ||
| XM_017009737.1 | XP_016865226.1 | 4 - 5 | 505 | 100 | ||
| XM_017009738.1 | XP_016865227.1 | 5 - 6 | 492 | 100 | ||
| XM_017009739.1 | XP_016865228.1 | 5 - 6 | 492 | 100 | ||
| XM_017009740.1 | XP_016865229.1 | 5 - 6 | 492 | 100 | ||
| XM_017009741.1 | XP_016865230.1 | 5 - 6 | 492 | 100 | ||
| XM_017009742.1 | XP_016865231.1 | 5 - 6 | 492 | 100 | ||
| XM_017009743.1 | XP_016865232.1 | 5 - 6 | 666 | 100 | ||
| XM_017009744.1 | XP_016865233.1 | 5 - 6 | 492 | 100 | ||
| XM_017009746.1 | XP_016865235.1 | 5 - 6 | 492 | 100 | ||
| GenBank mRNA | AF222747.1 | - | 5 - 6 | 492 | 100 | |
| AJ271459.1 | - | 5 - 6 | 500 | 100 | ||
| AJ288952.1 | - | 5 - 6 | 500 | 100 | ||
| AJ288953.1 | - | 5 - 6 | 500 | 100 | ||
| AJ288954.1 | - | 5 - 6 | 500 | 100 | ||
| AJ288955.1 | - | 5 - 6 | 500 | 100 | ||
| AK027880.1 | - | 3 - 4 | 320 | 100 | ||
| AK298785.1 | - | 5 - 6 | 649 | 100 | ||
| AK315159.1 | - | 5 - 6 | 586 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000938.hg.1
Set Membership: |
|

RANBP17
Genomic Map