Assay Details
| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF159165.1 | - | 1 | 96 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF159165.1 transcripts.
Set Membership: |
|

FAXDC2
Genomic Map