| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
TC06001298.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
ENST00000259711.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
ENST00000378814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
ENST00000378816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
ENST00000378826.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
ENST00000378843.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
NM_001105566.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
NM_001105567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
NM_001105568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015577.hg.1 |
NM_022113.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000259711.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000358380.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000378814.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000378826.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000378843.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000502297.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000506044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000514714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
NM_001105566.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
NM_001105567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
NM_001105568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
NM_022113.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000259711.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000358380.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000378814.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000378826.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000378843.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000502297.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000506044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
ENST00000514714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
NM_001105566.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
NM_001105567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
NM_001105568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
NM_022113.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600174307.hg.1 |
TC0600010954.hg.1 |
Clariom D TC |