Assay Details
| Gene Symbol: TWF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007284.3 | NP_009215.1 | 3 - 4 | 434 | 51 | |
| GenBank mRNA | AF109365.1 | - | 3 - 4 | 335 | 51 | |
| AL136773.1 | - | 3 - 4 | 401 | 51 | ||
| BC000327.2 | - | 3 - 4 | 385 | 2822004 | 51 | |
| BC003161.1 | - | 3 - 4 | 351 | 2957960 | 51 | |
| BC016452.1 | - | 3 - 4 | 401 | 3452300 | 51 | |
| CR456732.1 | - | 3 - 4 | 283 | 51 | ||
| CR533520.1 | - | 3 - 4 | 283 | 51 | ||
| Y17169.1 | - | 3 - 4 | 387 | 51 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001448.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of lamellipodium assembly
positive regulation of neuron projection development
cell projection organization
negative regulation of actin filament polymerization
regulation of microvillus length
regulation of actin cytoskeleton organization
sequestering of actin monomers
positive regulation of axon extension
barbed-end actin filament capping
cellular response to retinoic acid
cellular response to growth factor stimulus
cell-cell adhesion
actin monomer binding
protein kinase C binding
protein binding
ATP binding
phosphatidylinositol-4,5-bisphosphate binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
positive regulation of lamellipodium assembly
positive regulation of neuron projection development
cell projection organization
negative regulation of actin filament polymerization
regulation of microvillus length
regulation of actin cytoskeleton organization
sequestering of actin monomers
positive regulation of axon extension
barbed-end actin filament capping
cellular response to retinoic acid
cellular response to growth factor stimulus
cell-cell adhesion
actin monomer binding
protein kinase C binding
protein binding
ATP binding
phosphatidylinositol-4,5-bisphosphate binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03026189.hg.1 | TC03001448.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026189.hg.1 | ENST00000305533.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026189.hg.1 | ENST00000494383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026189.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026189.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026190.hg.1 | TC03001448.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026190.hg.1 | ENST00000305533.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026190.hg.1 | ENST00000494383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026190.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026190.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026192.hg.1 | TC03001448.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026192.hg.1 | ENST00000305533.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026192.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026192.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026191.hg.1 | TC03001448.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026191.hg.1 | ENST00000305533.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026191.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026191.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207933.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207933.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207933.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207933.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207933.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207933.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207933.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207933.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207933.hg.1 | TC0300013991.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300207935.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207935.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207935.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207935.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207935.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207935.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207935.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207935.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207935.hg.1 | TC0300013991.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300207934.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207934.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207934.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207934.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207934.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207934.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207934.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207934.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207934.hg.1 | TC0300013991.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000305533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000472755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | ENST00000499914.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013991.hg.1 | NM_007284.3 | Refseq |

TWF2
Genomic Map