Assay Details
| Gene Symbol: AKAP8L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291478.1 | NP_001278407.1 | 9 - 10 | 1178 | 58 | |
| NM_014371.3 | NP_055186.3 | 9 - 10 | 1361 | 58 | ||
| XM_005259854.3 | XP_005259911.2 | 10 - 11 | 1166 | 58 | ||
| XM_011527916.2 | XP_011526218.1 | 9 - 10 | 1268 | 58 | ||
| GenBank mRNA | AB025905.1 | - | 9 - 10 | 1208 | 58 | |
| AB451326.1 | - | 8 - 9 | 1163 | 58 | ||
| AB451469.1 | - | 8 - 9 | 1163 | 58 | ||
| AF199414.1 | - | 8 - 9 | 1163 | 58 | ||
| AJ243467.1 | - | 9 - 10 | 1209 | 58 | ||
| AK225046.1 | - | 9 - 10 | 1228 | 58 | ||
| AK225291.1 | - | 9 - 10 | 1228 | 58 | ||
| AK295956.1 | - | 9 - 10 | 1083 | 58 | ||
| BC000713.2 | - | 9 - 10 | 1213 | 3504388 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

AKAP8L
Genomic Map