Assay Details
| Gene Symbol: CRLF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004750.4 | NP_004741.1 | 5 - 6 | 1056 | 59 | |
| XM_011528422.2 | XP_011526724.1 | 5 - 6 | 1138 | 59 | ||
| XM_011528423.2 | XP_011526725.1 | 5 - 6 | 862 | 59 | ||
| XM_011528424.2 | XP_011526726.1 | 5 - 6 | 1130 | 59 | ||
| GenBank mRNA | AF059293.1 | - | 5 - 6 | 980 | 59 | |
| AF073515.1 | - | 5 - 6 | 1065 | 59 | ||
| AF178684.1 | - | 5 - 6 | 913 | 59 | ||
| AY358291.1 | - | 5 - 6 | 992 | 59 | ||
| BC044634.1 | - | 5 - 6 | 989 | 5926400 | 59 | |
More Information
Set Membership: |
|

CRLF1
Genomic Map