Assay Details
| Gene Symbol: TSPAN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168320.1 | NP_001161792.1 | 6 - 7 | 679 | 57 | |
| NM_006675.4 | NP_006666.1 | 7 - 8 | 746 | 57 | ||
| XM_011520912.2 | XP_011519214.1 | 8 - 9 | 863 | 57 | ||
| XM_011520913.2 | XP_011519215.1 | 5 - 6 | 605 | 57 | ||
| GenBank mRNA | AF089749.1 | - | 7 - 8 | 689 | 57 | |
| AK027366.1 | - | 7 - 8 | 722 | 57 | ||
| AK223179.1 | - | 7 - 8 | 718 | 57 | ||
| BC071881.1 | - | 6 - 7 | 661 | 6256950 | 57 | |
| CR542109.1 | - | 5 - 6 | 563 | 57 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000036.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12000503.hg.1 | ENST00000011898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000503.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000503.hg.1 | ENST00000537971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000503.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000503.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000503.hg.1 | TC12000036.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000506.hg.1 | ENST00000011898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000506.hg.1 | ENST00000537971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000506.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000506.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000506.hg.1 | TC12000036.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000505.hg.1 | ENST00000011898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000505.hg.1 | ENST00000537971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000505.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000505.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000505.hg.1 | TC12000036.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200144915.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144915.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144915.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144915.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144915.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144915.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144915.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144915.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144915.hg.1 | TC1200006520.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144914.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144914.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144914.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144914.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144914.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144914.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144914.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144914.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144914.hg.1 | TC1200006520.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144911.hg.1 | TC1200006520.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | ENST00000407263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144912.hg.1 | TC1200006520.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200144916.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144916.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144916.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144916.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144916.hg.1 | ENST00000011898.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144916.hg.1 | ENST00000537971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144916.hg.1 | NM_001168320.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144916.hg.1 | NM_006675.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144916.hg.1 | TC1200006520.hg.1 | Clariom D TC |

TSPAN9
Genomic Map