Assay Details
| Gene Symbol: ZBTB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317990.1 | NP_001304919.1 | 3 | 2913 | 68 | |
| NM_015898.3 | NP_056982.1 | 3 | 3077 | 68 | ||
| XM_005259570.4 | XP_005259627.3 | 3 | 3999 | 68 | ||
| XM_005259571.4 | XP_005259628.1 | 3 | 3847 | 68 | ||
| GenBank mRNA | CB047382.1 | - | 1 | 60 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

ZBTB7A
Genomic Map