| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02015519.hg.1 |
TC02001035.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02015519.hg.1 |
ENST00000306721.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02015519.hg.1 |
ENST00000347703.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02015519.hg.1 |
ENST00000392567.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02015519.hg.1 |
ENST00000410019.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02015519.hg.1 |
ENST00000410101.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02015519.hg.1 |
NM_031942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02015519.hg.1 |
NM_145810.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000306721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000347703.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000410019.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000410101.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000467411.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000496441.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
NM_031942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
NM_145810.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000306721.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000347703.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000410019.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000410101.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000467411.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
ENST00000496441.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
NM_031942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
NM_145810.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200170258.hg.1 |
TC0200009994.hg.1 |
Clariom D TC |