Assay Details
| Gene Symbol: RBM45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152945.3 | NP_694453.2 | 8 - 9 | 1358 | 118 | |
| XM_005246287.4 | XP_005246344.1 | 8 - 9 | 1374 | 118 | ||
| XM_017003320.1 | XP_016858809.1 | 8 - 9 | 1801 | 118 | ||
| XM_017003321.1 | XP_016858810.1 | 8 - 9 | 1802 | 118 | ||
| XM_017003322.1 | XP_016858811.1 | 8 - 9 | 1367 | 118 | ||
| GenBank mRNA | AB036991.1 | - | 8 - 9 | 1286 | 118 | |
| AF526533.1 | - | 7 - 8 | 1534 | 118 | ||
| AK126575.1 | - | 8 - 9 | 1216 | 118 | ||
| BC066549.1 | - | 8 - 9 | 1327 | 5748065 | 118 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001069.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02015875.hg.1 | TC02001069.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015875.hg.1 | ENST00000286070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015875.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000464647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000464647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000464647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000464647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000464647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000464647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000493048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010112.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000286070.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000424000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000424099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000455903.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | ENST00000616198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | NM_152945.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200170829.hg.1 | TC0200010112.hg.1 | Clariom D TC |

RBM45
Genomic Map