| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003678.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003678.hg.1 |
ENST00000328842.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003678.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003678.hg.1 |
TC22000198.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200149076.hg.1 |
TC2200007023.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200149076.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200149076.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200149076.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200149076.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
ENST00000310624.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007023.hg.1 |
NM_021076.3 |
Refseq |