Assay Details
| Gene Symbol: C22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057349.1 | - | 1 - 2 | 250 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK057349.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |

C22orf23
Genomic Map