Assay Details
| Gene Symbol: C22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032561.4 | NP_115950.3 | 3 - 4 | 380 | 64 | |
| XM_005261781.1 | XP_005261838.1 | 3 - 4 | 344 | 64 | ||
| XM_005261782.3 | XP_005261839.1 | 3 - 4 | 224 | 64 | ||
| GenBank mRNA | AF324466.1 | - | 2 - 3 | 180 | 64 | |
| AK057349.1 | - | 3 - 4 | 430 | 64 | ||
| BC031998.1 | - | 3 - 4 | 262 | 4755328 | 64 | |
| BI463610.1 | - | 3 - 4 | 385 | 64 | ||
| CR456440.1 | - | 3 - 4 | 260 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000780.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22013030.hg.1 | ENST00000249079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013030.hg.1 | ENST00000403026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013030.hg.1 | ENST00000403305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013030.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013030.hg.1 | TC22000780.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013029.hg.1 | ENST00000249079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013029.hg.1 | ENST00000403026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013029.hg.1 | ENST00000403305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013029.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013029.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22013029.hg.1 | TC22000780.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | TC2200008675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162666.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | TC2200008675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162665.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000249079.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403026.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000403305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000418863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000422191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | ENST00000619227.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_001207062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008675.hg.1 | NM_032561.4 | Refseq |

C22orf23
Genomic Map