Assay Details
| Gene Symbol: GM140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046249.1 | 1 | 57 | 99 | ||
| GenBank mRNA | AF387616.1 | - | 1 | 57 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928973 | LOC101928973 | -54.2kb | Sense | Chr. 1 - 181174484 - 181182208 on NCBI Build GRCh38 | |
| Downstream | calcium voltage-gated channel subunit alpha1 E | CACNA1E |
BII, CACH6, CACNL1A6, Cav2.3, gm139 |
246.8kb | Sense | Chr. 1 - 181317712 - 181808084 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696853 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696863 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637356 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637379 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696872 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637360 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637368 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637364 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637366 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696842 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637365 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637389 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696838 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637378 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696846 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00637363 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20696870 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3227440 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14016 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3227440 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P35378937 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P30392847 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35378937 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7907908 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7907909 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7907910 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7907907 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010821.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010821.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010821.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010821.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010821.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010821.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010821.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010821.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179522.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179522.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179522.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179522.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179522.hg.1 | TC0100010821.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010821.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010821.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010821.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010821.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010821.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010821.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010821.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010821.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179521.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179521.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179521.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179521.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179521.hg.1 | TC0100010821.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100179523.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179523.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179523.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179523.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179523.hg.1 | TC0100010821.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100179520.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179520.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179520.hg.1 | ENST00000319701.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179520.hg.1 | NR_046249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179520.hg.1 | TC0100010821.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 228628 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina human-6 v2.0 expression beadchip | ILMN_1879239 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_99443 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_99443 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1879239 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1879239 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1879239 | - | - |

GM140
Genomic Map