Assay Details
| Gene Symbol: SORBS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034956.1 | NP_001030128.1 | 16 - 17 | 1510 | 117 | |
| GenBank mRNA | AF356527.1 | - | 16 - 17 | 1521 | 117 | |
More Information
Set Membership: |
|

SORBS1
Genomic Map