Assay Details
| Gene Symbol: RANBP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017009747.1 | XP_016865236.1 | 1 - 2 | 128 | 104 | |
| GenBank mRNA | AJ288952.1 | - | 16 - 17 | 2003 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017009747.1 transcripts.
Set Membership: |
|

RANBP17
Genomic Map